missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NTE Polyclonal Antibody
GREENER_CHOICE

Product Code. 15685025
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15685025 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15685025 Supplier Invitrogen™ Supplier No. PA551106

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NTE Polyclonal Antibody detects endogenous levels of NTE protein.

This gene encodes a phospholipase that deacetylates intracellular phosphatidylcholine to produce glycerophosphocholine. It is thought to function in neurite outgrowth and process elongation during neuronal differentiation. The protein is anchored to the cytoplasmic face of the endoplasmic reticulum in both neurons and non-neuronal cells. Mutations in this gene result in autosomal recessive spastic paraplegia, and the protein is the target for neurodegeneration induced by organophosphorus compounds and chemical warfare agents. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTE
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene pnpla6
Gene Accession No. Q3TRM4, Q8IY17
Gene Alias 5'-NT; 5'-nucleotidase; 5'-nucleotidase ecto; 5'-nucleotidase, ecto (CD73); AI661849; BNHS; CALJA; CD73; E5NT; ecto-5'-nucleotidase; eN; eNT; iPLA2delta; LNMS; MSws; Neuropathy target esterase; NT; NT5; NT5E; NTE; NTEMND; OMCS; patatin like phospholipase domain containing 6; patatin-like phospholipase domain containing 6; patatin-like phospholipase domain-containing protein 6; PNPLA6; Purine 5-Prime-Nucleotidase; RGD1564611; SPG39; Swiss cheese; Swiss-cheese; sws; wu:fy20f06
Gene Symbols pnpla6
Host Species Rabbit
Immunogen Synthesized peptide derived from the Internal region of human NTE.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10908, 360753, 50767
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.