missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Buffer NTC

Product Code. 11922312
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11922312 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11922312 Supplier Macherey-Nagel™ Supplier No. 740654.100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Binding Buffer NTC to be used with NucleoSpin Gel and PCR Clean‐up kit; for clean up of single stranded DNA

NucleoSpin™ Gel and PCR Clean-Up is a two-in-one kit with optimized recovery and elution volume for both PCR purification and gel extraction. The protocol ensures complete removal of contaminations for use in downstream applications, such as cloning, sequencing, PCR and restriction analysis.

Binding Buffer NTC is a supplemental buffer that obtains higher yields for small fragments compared to Buffer NTI, which is included in the kit. Generally, 200 μL Buffer NTC is mixed with 100 μL sample or 100 mg agarose gel <2%. (Use 400 μL NTC for 100 mg agarose gel >2%.)

Applications:

  • Clean-up/purification of short, single-stranded DNA
  • Extract small, single-stranded RNA from agarose gels
  • Achieve full recovery of fragments <50bp, even primer oligonucleotides

Specifications

Chemical Name or Material Binding Buffer NTC
For Use With (Application) For clean-up of single-stranded DNA and for Extraction of single-stranded RNA from agarose gel
For Use With (Equipment) NucleoSpin™ Gel and PCR Clean-Up Kit
Quantity 100 mL
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
Supplemental information
  • MIXTURE LIST-Contain: potassium thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.