missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Binding Buffer NTB
Description
NucleoTrap™ is a silica matrix based kit for isolation of highly pure DNA fragments from agarose gels and aqueous solutions (desalination). The kit can also be used for the removal of nucleotides and/or biotin after labelling reactions as well as for the removal of enzymes. By means of a specially activated spherical silica matrix it is possible to extract DNA fragments from agarose gels or solvents in a very mild manner, without the use of organic solvents. Nucleic acids are bound to the NucleoTrap™ matrix in the presence of a chaotropic salt. The hydrate shell of the nucleic acids is destroyed by this binding buffer, thus facilitating adsorption to the specially treated NucleoTrap™ matrix. A washing step is followed by the elution of DNA with low-salt elution buffer or distilled water. Kit components: NucleoTrap™ suspension, buffers.
- Silica matrix technology
- Fragment size: 20bp to 20kb
- Sample material: 20mg to 600mg agarose gel
- Elution volume: 20μL to 50μL
- Binding capacity: 6μg/10μL suspension
- Typical Recovery: 50% to 90%
- Preparation time: 60 min./6 preps
- Format: silica bead suspension For extraction of DNA from agarose gels, suitable, for example, for subsequent cloning sequencing, PCR, restriction analysis. Support protocol for concentration and desalination of reaction mixtures.
Specifications
Specifications
| Chemical Name or Material | Wash Buffer NTB |
| For Use With (Application) | For Clean-up |
| Quantity | 1000 mL |
Safety and Handling
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?