missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NTB-A/SLAMF6/CD352 Antibody (SGN-CD352A) - Humanized, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28645-100ug
This item is not returnable.
View return policy
Description
NTB-A/SLAMF6/CD352 Monoclonal antibody specifically detects NTB-A/SLAMF6/CD352 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| NTB-A/SLAMF6/CD352 | |
| Monoclonal | |
| Unconjugated | |
| Q96DU3 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| SGN-CD352A | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Activating NK receptor, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6 | |
| SLAMF6/ CD352 | |
| 100 μg | |
| Immunology | |
| 114836 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction