missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTB-A/SLAMF6/CD352 Antibody (292811), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB19081MFV450
420.75 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
NTB-A/SLAMF6/CD352 Monoclonal antibody specifically detects NTB-A/SLAMF6/CD352 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| NTB-A/SLAMF6/CD352 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Immunology | |
| 114836 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 292811 | |
| Flow Cytometry, CyTOF-ready | |
| Activating NK receptor, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6 | |
| Mouse myeloma cell line NS0-derived recombinant human NTB-A/SLAMF6, Leu28-Lys225, Accession # Q96DU3 | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction