missing translation for 'onlineSavingsMsg'
Learn More

NTB-A/SLAMF6/CD352 Antibody (292811), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals FAB19081M

440.30 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


Product Code. 18510859

  • £542.00 / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NTB-A/SLAMF6/CD352 Monoclonal antibody specifically detects NTB-A/SLAMF6/CD352 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NTB-A/SLAMF6/CD352
Monoclonal
DyLight 594
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, CyTOF
292811
Flow Cytometry, CyTOF-ready
Activating NK receptor, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6
Mouse myeloma cell line NS0-derived recombinant human NTB-A/SLAMF6, Leu28-Lys225, Accession # Q96DU3
0.1 mL
Immunology
114836
Store at 4°C in the dark.
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.