missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTB-A/SLAMF6/CD352 Antibody (002), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90026FR
This item is not returnable.
View return policy
Description
NTB-A/SLAMF6/CD352 Monoclonal specifically detects NTB-A/SLAMF6/CD352 in Human samples. It is validated for Flow Cytometry.
Specifications
| NTB-A/SLAMF6/CD352 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 2 | |
| Flow Cytometry | |
| Activating NK receptor, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human NTB-A/SLAMF6/CD352 (Q96DU3-1; Met1-Met226). | |
| 0.1 mL | |
| Immunology | |
| 114836 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction