missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTAL Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£161.00 - £408.00
Specifications
| Antigen | NTAL |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17939662
![]() |
Invitrogen™
PA5145377 |
200 μL |
£408.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17969462
|
Invitrogen™
PA5145378 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NTAL (Non-T cell Activation Linker), also known as LAB (Linker for activation of B cells), is a 30 kDa double-palmitoylated transmembrane adaptor protein expressed by B cells, NK cells, mast cells and macrophages. It is a negative regulator of early stages of BCR-dependent B cell signaling and serves as a negative regulator also in mast cells. However, in mast cells, NTAL also contributes to some activation processes, partially overlapping with LAT function.Specifications
| NTAL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q8CGL2, Q9GZY6, Q9JHL0 | |
| 317676, 56743, 7462 | |
| Recombinant Rat LAT2 protein, Arg32-Asp192 (Accession #Q8CGL2). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| AW125574; HSPC046; LAB; Lat2; LAT-2 like; Linker for activation of B-cells; linker for activation of T cells family, member 2; linker for activation of T cells, transmembrane adaptor 2; linker for activation of T-cells family member 2; Membrane-associated adapter molecule; non-T cell activation linker; non-T-cell activation linker; NTAL; WBS15; WBSCR15; WBSCR5; Williams-Beuren syndrome chromosomal region 15 protein; williams-Beuren syndrome chromosomal region 15 protein homolog; Williams-Beuren syndrome chromosomal region 5 protein; Williams-Beuren syndrome chromosome region 5 homolog; Williams-Beuren syndrome chromosome region 5-like protein; WSCR5 | |
| LAT2 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| LAT2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title