Learn More
Invitrogen™ NTAL Monoclonal Antibody (NAP-03)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA119327
Description
This antibody recognizes a defined intracellular epitope (aa 196-212) of human Non-T cell Activation Linker (NTAL), also known as LAB (linker of activated B cells), a 30 kDa transmembrane adaptor protein associating with lipid raft components in the plasma membrane of B cells, NK cells and myeloid cells. This antibody will not cross-react with mouse. Western Blot: Incubation 60 min; non-reducing conditions, 12% separating SDS-PAGE gel.
NTAL (Non-T cell Activation Linker), also known as LAB (Linker for activation of B cells), is a 30 kDa double-palmitoylated transmembrane adaptor protein expressed by B cells, NK cells, mast cells and macrophages. It is a negative regulator of early stages of BCR-dependent B cell signaling and serves as a negative regulator also in mast cells. However, in mast cells, NTAL also contributes to some activation processes, partially overlapping with LAT function.
Specifications
| NTAL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 15 mM sodium azide; pH 7.4 | |
| Q9GZY6 | |
| LAT2 | |
| Peptide corresponding to aa 196-212 of human NTAL, coupled to KLH. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| NAP-03 | |
| Unconjugated | |
| LAT2 | |
| AW125574; HSPC046; LAB; Lat2; LAT-2 like; Linker for activation of B-cells; linker for activation of T cells family, member 2; linker for activation of T cells, transmembrane adaptor 2; linker for activation of T-cells family member 2; Membrane-associated adapter molecule; non-T cell activation linker; non-T-cell activation linker; NTAL; WBS15; WBSCR15; WBSCR5; Williams-Beuren syndrome chromosomal region 15 protein; williams-Beuren syndrome chromosomal region 15 protein homolog; Williams-Beuren syndrome chromosomal region 5 protein; Williams-Beuren syndrome chromosome region 5 homolog; Williams-Beuren syndrome chromosome region 5-like protein; WSCR5 | |
| Mouse | |
| Protein A | |
| RUO | |
| 7462 | |
| 4°C, do not freeze | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.