missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTAL Antibody (NAP-07), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-341PCP
This item is not returnable.
View return policy
Description
NTAL Monoclonal specifically detects NTAL in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| NTAL | |
| Monoclonal | |
| PerCP | |
| LABWilliams-Beuren syndrome chromosomal region 15 protein, Linker for activation of B-cells, linker for activation of T cells family, member 2, linker for activation of T-cells family member 2, Membrane-associated adapter molecule, Non-T-cell activation linker, NTALWilliams-Beuren syndrome chromosomal region 5 protein, WBS15, WBSCR15, WBSCR5HSPC046, Williams-Beuren syndrome chromosome region 5, WSCR5 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody NAP-07 reacts with Non-T cell activation linker (NTAL), also known as LAB (linker of activated B cells), a 25 - 30 kDa transmembrane adaptor protein present in membrane microdomains (rafts) of B lymphocytes, NK cells and myeloid cells. | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| NAP-07 | |
| Flow Cytometry | |
| LAT2 | |
| Recombinant cytoplasmic domain (amino acids 91-244) of Human NTAL. | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Immunology, Innate Immunity, Signal Transduction, Transcription Factors and Regulators | |
| 7462 | |
| Human, Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction