missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NT5C3B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563399
This item is not returnable.
View return policy
Description
Immunogen sequence: IVSNYMDFNE DGFLQGFKGQ LIHTYNKNSS VCENCGYFQQ LEGKTNVILL GDSIG Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
Specifically hydrolyzes 7-methylguanosine monophosphate (m(7)GMP) to 7-methylguanosine and inorganic phosphate (PubMed:23223233, PubMed:24603684). The specific activity for m(7)GMP may protect cells against undesired salvage of m(7)GMP and its incorporation into nucleic acids (PubMed:23223233). Also has weak activity for CMP (PubMed:23223233, PubMed:24603684). UMP and purine nucleotides are poor substrates (PubMed:23223233). [UniProt].
Specifications
| NT5C3B | |
| Polyclonal | |
| Unconjugated | |
| NT5C3B | |
| 2610037D24Rik; 5'-nucleotidase, cytosolic IIIB; 5'-nucleotidase, cytosolic III-like; 7-methylguanosine nucleotidase; 7-methylguanosine phosphate-specific 5'-nucleotidase; AI841843; C330027I04Rik; cN-IIIB; cN-III-like protein; Cytosolic 5'-nucleotidase 3B; cytosolic 5'-nucleotidase III-like protein; cytosolic 5-nucleotidase III-like protein-like; N(7)-methylguanylate 5'-phosphatase; Nt5c3b; Nt5c3l | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 115024 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q969T7 | |
| NT5C3B | |
| Recombinant protein corresponding to Human NT5C3B. Recombinant protein control fragment (Product #RP-103290). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction