missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NT5C3A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144692
This item is not returnable.
View return policy
Description
Immunogen sequence: FDKLQQHSIP VFIFSAGIGD VLEEVIRQAG VYHPNVKVVS NFMDFDETGV LKGFKGELIH VFNKHDGALR NTEYFNQLKD NSNIILLGDS QGDLRMADGV ANVEHILKIG YLNDRVDELL EKYMDSYDIV LVQDESLEVA NSILQKI Antigen sequence identity: Mouse 93%; Rat 93%.
Pyrimidine 5-prime-nucleotidase (P5N), also called uridine 5-prime monophosphate hydrolase (UMPH), catalyzes the dephosphorylation of the pyrimidine 5-prime monophosphates UMP and CMP to the corresponding nucleosides. There are 2 isozymes of pyrimidine 5-prime nucleotidase in red blood cells, referred to as type I (UMPH1) and type II (UMPH2). The 2 enzymes are not separable by electrophoresis in humans but have distinct kinetic properties, and the proteins show no homology.
Specifications
| NT5C3A | |
| Polyclonal | |
| Unconjugated | |
| NT5C3A | |
| 1600024P05Rik; 2610206B05Rik; 3110004A18Rik; 5'-nucleotidase, cytosolic III; 5'-nucleotidase, cytosolic IIIA; 5'-nucleotidase, pyrimidine; 7-methylguanosine nucleotidase; 7-methylguanosine phosphate-specific 5'-nucleotidase; AI842776; cN-III; Cytosolic 5'-nucleotidase 3; cytosolic 5'-nucleotidase 3A; Cytosolic 5'-nucleotidase III; fb30h07; HSPC233; hUMP1; lupin; MGC27337; MGC87109; MGC87828; nt5c3; Nt5c3a; Nt5y; p36; P5N1; P5'N-1; P5N-1; PN-1; PN-I; POMP; PSN1; pyrimidine 5'-nucleotidase; pyrimidine 5'-nucleotidase 1; UMPH; UMPH1; Umph-1; Unknown (protein for MGC:133762); uridine 5'-monophosphate hydrolase 1; uridine monophosphatase 1; wu:fb30h07; zgc:66117 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51251 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H0P0 | |
| NT5C3A | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 189-335. Recombinant protein control fragment (Product #RP-102536). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction