missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT5C3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11393-1-AP-150UL
This item is not returnable.
View return policy
Description
Pyrimidine 5-prime-nucleotidase (P5N), also called uridine 5-prime monophosphate hydrolase (UMPH), catalyzes the dephosphorylation of the pyrimidine 5-prime monophosphates UMP and CMP to the corresponding nucleosides. There are 2 isozymes of pyrimidine 5-prime nucleotidase in red blood cells, referred to as type I (UMPH1) and type II (UMPH2). The 2 enzymes are not separable by electrophoresis in humans but have distinct kinetic properties, and the proteins show no homology.Specifications
| NT5C3 | |
| Polyclonal | |
| Unconjugated | |
| NT5C3A | |
| 5′ nucleotidase, cytosolic III, cN III, Cytosolic 5 nucleotidase 3, Cytosolic 5 nucleotidase III, NT5C3, p36, P5N 1, P5′N 1, P5N1, PN I, PSN1, Pyrimidine 5 nucleotidase 1, UMPH, UMPH1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 107569, 312373, 51251 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9H0P0 | |
| Nt5c3, Nt5c3a | |
| NT5C3 Fusion Protein Ag1949 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction