missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT5C3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93332-0.02ml
This item is not returnable.
View return policy
Description
NT5C3 Polyclonal antibody specifically detects NT5C3 in Human, Mouse samples. It is validated for Western Blot
Specifications
| NT5C3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| 5'-nucleotidase, cytosolic III, cN-IIIMGC87828, cytosolic 5'-nucleotidase 3, Cytosolic 5'-nucleotidase III, p36, P5N1, P5N-1, P5'N-1MGC27337, PN-IMGC87109, PSN1UMPH, Pyrimidine 5'-nucleotidase 1, UMPH1EC 3.1.3.5, uridine 5' monophosphate hydrolase 1, Uridine 5'-monophosphate hydrolase 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 37-336 of human NT5C3A (NP_001002010.1). TLKRKTGRKTKIIEMMPEFQKSSVRIKNPTRVEEIICGLIKGGAAKLQIITDFDMTLSRFSYKGKRCPTCHNIIDNCKLVTDECRKKLLQLKEKYYAIEVDPVLTVEEKYPYMVEWYTKSHGLLVQQALPKAKLKEIVAESDVMLKEGYENFFDKLQQHSIPVFIFSAGIGDVLEEVIRQAGVYHPNVKVVSNFMDFDETGVLKGFKGELIHVFNKHDGALRNTEYFNQLKDNSNIILLGDSQGDLRMADGVANVEHILKIGYLNDRVDELLEKYMDSYDIVLVQDESLEVANSILQKIL | |
| 0.02 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51251 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction