missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT5C3 Antibody (009), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90252FR
This item is not returnable.
View return policy
Description
NT5C3 Monoclonal specifically detects NT5C3 in Human samples. It is validated for Western Blot.
Specifications
| NT5C3 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 51251 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| 9 | |
| Western Blot | |
| 5'-nucleotidase, cytosolic III, cN-IIIMGC87828, cytosolic 5'-nucleotidase 3, Cytosolic 5'-nucleotidase III, p36, P5N1, P5N-1, P5'N-1MGC27337, PN-IMGC87109, PSN1UMPH, Pyrimidine 5'-nucleotidase 1, UMPH1EC 3.1.3.5, uridine 5' monophosphate hydrolase 1, Uridine 5'-monophosphate hydrolase 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human NT5C3 (NP_001002010; Met12-Leu297). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction