missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT5C2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31404
This item is not returnable.
View return policy
Description
NT5C2 Polyclonal specifically detects NT5C2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| NT5C2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| NT5C2 | |
| Recombinant protein encompassing a sequence within the center region of human NT5C2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| 5'-nucleotidase (purine), cytosolic type B, 5'-nucleotidase, cytosolic II, cN-II, Cytosolic 5'-nucleotidase II, EC 3.1.3, EC 3.1.3.5, GMP, NT5BIMP-specific 5'-NT, NT5CP, PNT5cytosolic purine 5'-nucleotidase, purine 5' nucleotidase | |
| Rabbit | |
| 65 kDa | |
| 100 μL | |
| Proteases & Other Enzymes | |
| 22978 | |
| Human, Mouse | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto