missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NT5C1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563340
This item is not returnable.
View return policy
Description
Immunogen sequence: SQTSLKQKKN EPGMRSSKES LEAEKRKESD KTGVRLSNQM RRAVNPNHSL RCCPFQGHSS CRRCLCAAEG TALGPCHTIR IYIHMCLLWE QG Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 77%.
Cytosolic 5-prime nucleotidases, such as NT5C1B, catalyze production of adenosine, which regulates diverse physiologic processes.
Specifications
| NT5C1B | |
| Polyclonal | |
| Unconjugated | |
| NT5C1B | |
| 4921514H13Rik; 5'nucleotidase; 5'-nucleotidase, cytosolic IB; AIRP; Autoimmune infertility-related protein; CN1B; cN-IB; cytosolic 5'-nucleotidase 1B; Cytosolic 5'-nucleotidase IB; FKSG85; NT5C1B; testicular tissue protein Li 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 93034 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96P26 | |
| NT5C1B | |
| Recombinant protein corresponding to Human NT5C1B. Recombinant protein control fragment (Product #RP-100561). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction