missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NT5C1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562933
This item is not returnable.
View return policy
Description
Immunogen sequence: DQMFHVAGAQ EMGTVAAHVP YGVAQTPRRT APAKQA Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 78%.
Dephosphorylates the 5' and 2'(3')-phosphates of deoxyribonucleotides and has a broad substrate specificity. Helps to regulate adenosine levels in heart during ischemia and hypoxia.
Specifications
| NT5C1A | |
| Polyclonal | |
| Unconjugated | |
| NT5C1A | |
| 5'-nucleotidase, cytosolic IA; AMP-specific 5'-NT; CN1; CN1A; CNI; CN-I; cN-IA; cytosolic 5' nucleotidase, type 1A; cytosolic 5'-nucleotidase 1A; Cytosolic 5'-nucleotidase IA; Gm49; NT5C1A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84618 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BXI3 | |
| NT5C1A | |
| Recombinant protein corresponding to Human NT5C1A. Recombinant protein control fragment (Product #RP-101220). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction