Learn More
NT5C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554851
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VLADFEAGLL RGFRRRFPEE PHVPLEQRRG FLAREQYRAL RPDLADKVAS VYEAPGFFLD LEPIP Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 72%.
Pyrimidine 5-prime nucleotidase and type II (UMPH2).Specifications
NT5C | |
Polyclonal | |
Unconjugated | |
NT5C | |
5' nucleotidase, deoxy (pyrimidine), cytosolic type C; 5'(3')-deoxyribonucleotidase, cytosolic type; 5', 3'-nucleotidase, cytosolic; 5',3'-nucleotidase, cytosolic; 5,3-nucleotidase, cytosolic; cdN; Cytosolic 5',3'-pyrimidine nucleotidase; deoxy-5'-nucleotidase 1; Dnt; DNT1; dNT-1; epididymis luminal protein 74; HEL74; Nt5c; P5N2; PN-I; PN-II; Umph2; Umph-2; uridine 5'-monophosphate phosphohydrolase 2; uridine 5-monophosphate phosphohydrolase 2; uridine 5-prime monophosphate hydrolase 2; uridine monophosphatase 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
30833 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TCD5 | |
NT5C | |
Recombinant protein corresponding to Human NT5C | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only