missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NT5C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554461
This item is not returnable.
View return policy
Description
Immunogen sequence: GALDAVREMN DLPDTQVFIC TSPLLKYHHC VGEKYRWVEQ HLGPQFVERI ILTRDKTVVL Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.
Pyrimidine 5-prime nucleotidase and type II (UMPH2).
Specifications
| NT5C | |
| Polyclonal | |
| Unconjugated | |
| NT5C | |
| 5' nucleotidase, deoxy (pyrimidine), cytosolic type C; 5'(3')-deoxyribonucleotidase, cytosolic type; 5', 3'-nucleotidase, cytosolic; 5',3'-nucleotidase, cytosolic; 5,3-nucleotidase, cytosolic; cdN; Cytosolic 5',3'-pyrimidine nucleotidase; deoxy-5'-nucleotidase 1; Dnt; DNT1; dNT-1; epididymis luminal protein 74; HEL74; Nt5c; P5N2; PN-I; PN-II; Umph2; Umph-2; uridine 5'-monophosphate phosphohydrolase 2; uridine 5-monophosphate phosphohydrolase 2; uridine 5-prime monophosphate hydrolase 2; uridine monophosphatase 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100360582, 30833 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TCD5 | |
| NT5C | |
| Recombinant protein corresponding to Human NT5C. Recombinant protein control fragment (Product #RP-93232). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction