missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Buffer NT3 Concentrate

Product Code. 10767092
Change view
Click to view available options
Quantity:
20 mL
Unit Size:
20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10767092 20 mL 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10767092 Supplier Macherey-Nagel™ Supplier No. 740598

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Wash Buffer NT3 Concentrate, for preparation of Buffer NT3 to be used with NucleoSpin Gel and PCR Clean‐up kits, NucleoSpin Gel and PCR Clean Midi / Maxi kits, NucleoTrap, or NucleoTraPCR kits.

Washing buffer is used to remove contamination from DNA that has been bound to the column membrane matrix during DNA purification and isolation. This wash buffer is available in a convenient concentrated form. The wash buffer solution should be prepared before initiating the DNA isolation protocol.

  • For use with Macherey-Nagel™ DNA isolation/purification systems, including NucleoTrap™ and NucleoSpin™ kits
  • To prepare the wash buffer, add four parts ethanol to 1 part wash buffer concentrate
  • (20 mL concentrate=100 mL solution)

Specifications

Chemical Name or Material Wash Buffer NT3
For Use With (Equipment) DNA purification/isolation kits, including NucleoTrap™ and NucleoSpin™
Physical Form Concentrate
Quantity 20 mL
Solubility Information Soluble in Ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.