Learn More
Macherey-Nagel™ NT1 Binding Buffer
Brand: Macherey-Nagel™ 740596.100
Code : 86
Additional Details : CAS Number : 7601-89-0 Weight : 0.14500kg Transport : UN number : 3211 Chem class : 5.1 Pack group : III
Description
NucleoTrap™ is a silica matrix based kit for isolation of highly pure DNA fragments from agarose gels and aqueous solutions (desalination). The kit can also be used for the removal of nucleotides and/or biotin after labelling reactions as well as for the removal of enzymes. By means of a specially activated spherical silica matrix it is possible to extract DNA fragments from agarose gels or solvents in a very mild manner, without the use of organic solvents. Nucleic acids are bound to the NucleoTrap™ matrix in the presence of a chaotropic salt. The hydrate shell of the nucleic acids is destroyed by this binding buffer, thus facilitating adsorption to the specially treated NucleoTrap™ matrix. A washing step is followed by the elution of DNA with low-salt elution buffer or distilled water. Kit components: NucleoTrap™ suspension, buffers.- Silica matrix technology
- Fragment size: 20bp to 20kb
- Sample material: 20mg to 600mg agarose gel
- Elution volume: 20μL to 50μL
- Binding capacity: 6μg/10μL suspension
- Typical Recovery: 50% to 90%
- Preparation time: 60min/6 preps
- Format: silica bead suspension For extraction of DNA from agarose gels, suitable, for example, for subsequent cloning sequencing, PCR, restriction analysis. Support protocol for concentration and desalination of reaction mixtures.
Specifications
100 mL | |
For Clean-up | |
Binding Buffer NT1 |
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 4
- Oxidising liquid Category 2
Hazard Statement
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
Supplemental information
- MIXTURE LIST-Contains : sodium perchlorate