missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT-proBNP Antibody (11D1cc), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
£436.00
Specifications
| Antigen | NT-proBNP |
|---|---|
| Clone | 11D1cc |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 750 |
Description
NT-proBNP Monoclonal antibody specifically detects NT-proBNP in Human samples. It is validated for Western Blot, ELISASpecifications
| NT-proBNP | |
| Western Blot | |
| Alexa Fluor 750 | |
| Mouse | |
| Biologically Active Proteins, Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience, Neurotransmission | |
| 50mM Sodium Borate | |
| 4879 | |
| IgG1 | |
| Protein A purified |
| 11D1cc | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with synthetic NT-proBNP, corresponding to a.a.r. 28-45, conjugated with carrier protein. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title