missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT-4 Antibody (OTI2E8), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73072F
This item is not returnable.
View return policy
Description
NT-4 Monoclonal specifically detects NT-4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| NT-4 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| NTF4 | |
| Recombinant protein fragment corresponding to amino acids 81-210 of of human NT-4 (NP_006170) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2E8 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| GLC10, GLC1O, neurotrophic factor 4, neurotrophic factor 5, neurotrophin 4, neurotrophin 5 (neurotrophin 4/5), neurotrophin-4, neurotrophin-5, Neutrophic factor 4, NT4, NT-4, NT-4/5, NT-5, NTF5NT5 | |
| Mouse | |
| Affinity Purified | |
| Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Neuroscience | |
| 4909.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction