missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NSUN5 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA583781

Product Code. 16350735

  • £478.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MPSRQLEDPG AGTPSPVRLH ALAGFQQRAL CHALTFPSLQ RL.

NSUN5 is a member of the evolutionarily conserved NOL1/NOP2/Sun domain family. The encoded protein may function as a DNA methyltransferase in the nucleus. The gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NSUN5
Polyclonal
Unconjugated
Nsun5
9830109N13Rik; AI326939; FLJ10267; MGC986; NOL1; NOL1/NOP2/Sun domain family member 5; NOL1/NOP2/Sun domain family, member 5; NOL1R; NOL1-related protein; NOP2/Sun domain family, member 5; NOP2/Sun domain family, member 5A; NOP2/Sun RNA methyltransferase family member 5; Nsun5; NSUN5A; p120; p120(NOL1); Probable 28S rRNA (cytosine-C(5))-methyltransferase; putative methyltransferase NSUN5; RGD1309268; WBSCR20; Wbscr20a; Williams Beuren syndrome chromosome region 20A; Williams Beuren syndrome chromosome region 20A homolog; Williams-Beuren syndrome chromosomal region 20A protein; Williams-Beuren syndrome chromosomal region 20A protein homolog; Williams-Beuren syndrome critical region protein 20; Williams-Beuren syndrome critical region protein 20 copy A
Rabbit
Antigen Affinity Chromatography
RUO
55695
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunofluorescence
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96P11
Nsun5
Recombinant protein corresponding to Human NSUN5.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.