missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSUN3 Polyclonal Antibody

NSUN3 Polyclonal Antibody
Brand: Invitrogen PA566119
This item is not returnable.
View return policy
Description
NSUN3 Polyclonal Antibody for ICC/IF
Specifications
| NSUN3 | |
| Polyclonal | |
| Unconjugated | |
| NSUN3 | |
| 6720484A09Rik; AU022521; MST077; MSTP077; NOL1/NOP2/Sun domain family 3; NOL1/NOP2/Sun domain family member 3; NOL1/NOP2/Sun domain family member 3-like; NOL1/NOP2/Sun domain family, member 3; NOP2/Sun domain family, member 3; NOP2/Sun RNA methyltransferase 3; NOP2/Sun RNA methyltransferase family member 3; Nsun3; putative methyltransferase NSUN3; tRNA (cytosine(34)-C(5))-methyltransferase, mitochondrial; UG0651E06; Unknown (protein for MGC:142518) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 63899 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H649 | |
| NSUN3 | |
| Recombinant Human NSUN3. Recombinant protein control fragment (Product #RP-106809). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction