missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NSUN2 Recombinant Rabbit Monoclonal Antibody (PSH05-05)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556501
This item is not returnable.
View return policy
Description
NSUN2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), IP
NSUN2 encodes a methyltransferase that catalyzes the intron-dependent formation of 5-methylcytosine at C34 of tRNA-leu(CAA). Maturation of cytoplasmic tRNAs includes splicing of introns, which are located 1 nucleotide 3-prime from the anticodon in all intron-containing tRNA genes. In tRNA-leu(CAA), the first position of the anticodon, C34, is converted to 5-methylcytosine, a modification necessary to stabilize the anticodon-codon pairing and correctly translate the mRNA. Mutations of NSUN2 result in autosomal recessive 5 mental retardation (MRT5).
Specifications
| NSUN2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q08J23, Q1HFZ0 | |
| NSUN2 | |
| Recombinant protein within human NSUN2 aa 1-767. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| PSH05-05 | |
| Unconjugated | |
| NSUN2 | |
| 5-methycytoisine methyltransferase; D13Wsu123e; hTrm4; Misu; mRNA cytosine C(5)-methyltransferase; MRT5; Myc-induced SUN domain containing protein; myc-induced SUN domain-containing protein; Myc-induced SUN-domain-containing protein; NOL1/NOP2/Sun domain family 2; NOL1/NOP2/Sun domain family member 2; NOL1/NOP2/Sun domain family, member 2; NOP2/Sun domain family, member 2; NOP2/Sun RNA methyltransferase family member 2; NOP2/Sun RNA methyltransferase family, member 2; Nsun2; RGD1311954; RNA cytosine C(5)-methyltransferase NSUN2; SAKI; substrate of AIM1/Aurora kinase B; TRM4; tRNA (cytosine(34)-C(5))-methyltransferase; tRNA (cytosine-5-)-methyltransferase NSUN2; tRNA cytosine C(5)-methyltransferase; tRNA methyltransferase 4 homolog | |
| Rabbit | |
| Protein A | |
| RUO | |
| 0, 28114, 54888 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction