missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NSRP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553330
This item is not returnable.
View return policy
Description
Immunogen sequence: KREVGVQSSE RNQDRKESSP NSRAKDKFLD QERSNKMRNM AKDKERNQEK PSNSESSLGA KHRLTEEGQE KGKEQERPPE AVSKFAKRNN EETVM Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 66%.
RNA-binding protein that mediates pre-mRNA alternative splicing regulation.
Specifications
| NSRP1 | |
| Polyclonal | |
| Unconjugated | |
| NSRP1 | |
| 8030451K07; AI851076; AL022898; Ccdc55; coiled-coil domain containing 55; coiled-coil domain-containing protein 55; HSPC095; NSpr70; NSRP1; NSrp70; Nuclear speckle splicing regulatory protein 1; nuclear speckle-related protein 70; RGD1309863 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84081 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H0G5 | |
| NSRP1 | |
| Recombinant protein corresponding to Human NSRP1. Recombinant protein control fragment (Product #RP-90746). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction