missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NSRP1 Polyclonal Antibody
GREENER_CHOICE

Chicken Polyclonal Antibody

Brand:  Invitrogen™ PA572714

Product Code. 15931375

  • £442.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NSRP1 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

RNA-binding protein that mediates pre-mRNA alternative splicing regulation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NSRP1
Polyclonal
Unconjugated
NSRP1
8030451K07; AI851076; AL022898; Ccdc55; coiled-coil domain containing 55; coiled-coil domain-containing protein 55; HSPC095; NSpr70; NSRP1; NSrp70; Nuclear speckle splicing regulatory protein 1; nuclear speckle-related protein 70; RGD1309863
Chicken
Antigen Affinity Chromatography
RUO
237859, 303346, 84081
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.02% sodium azide
Q4FZU3, Q5NCR9, Q9H0G5
NSRP1
CCDC55 antibody for a 16 amino acid synthetic peptide near the carboxy terminus of human CCDC55. The immunogen is within amino acids 430 - 480 of CCDC55.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.