missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSRP1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94395-0.02ml
This item is not returnable.
View return policy
Description
NSRP1 Polyclonal antibody specifically detects NSRP1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NSRP1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CCDC55, coiled-coil domain containing 55, coiled-coil domain-containing protein 55, DKFZp434K1421, FLJ37233, HSPC095, NSRP70, nuclear speckle splicing regulatory protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human NSRP1 (NP_115517.1). MAIPGRQYGLILPKKTQQLHPVLQKPSVFGNDSDDDDETSVSESLQREAAKKQAMKQTKLEIQKALAEDATVYEYDSIYDEMQKKKEENNPKLLLGKDRKPKYIHNLLKAVEIRKKEQEKRMEKKIQREREMEKGEFDDKEAFVTSAYKKKLQERAEEEEREKRAAALEACLDVTKQKDLSGFYRHLLNQAVGEEEVPKCSFREARSGIKEEKSRGFSNEVSSKNRIPQEKCILQTDVKVEENPDADSDFDAKSSADDEIEETRVNCRREKVIETPENDF | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84081 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction