missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ nSMase Antibody (2C9), Novus Biologicals™

Product Code. 18343979 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343979 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18343979 Supplier Novus Biologicals™ Supplier No. H00006610M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

nSMase Monoclonal antibody specifically detects nSMase in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen nSMase
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Sandwich ELISA
Classification Monoclonal
Clone 2C9
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH00038
Gene Alias EC 3.1.4.12, ISC1, lyso-PAF-PLC, Lyso-platelet-activating factor-phospholipase C, Neutral sphingomyelinase, nSMase, N-SMase, NSMASE1, sphingomyelin phosphodiesterase 2, sphingomyelin phosphodiesterase 2, neutral membrane (neutral sphingomyelinase)
Host Species Mouse
Immunogen SMPD2 (AAH00038.1, 1 a.a. ∽ 423 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MKPNFSLRLRIFNLNCWGIPYLSKHRADRMRRLGDFLNQESFDLALLEEVWSEQDFQYLRQKLSPTYPAAHHFRSGIIGSGLCVFSKHPIQELTQHIYTLNGYPYMIHHGDWFSGKAVGLLVLHLSGMVLNAYVTHLHAEYNRQKDIYLAHRVAQAWELAQFIHHTSKKADVVLLCGDLNMHPEDLGCCLLKEWTGLHDAYLETRDFKGSEEGNTMVPKNCYVSQQELKPFPFGVRIDYVLYKAVSGFYISCKSFETTTGFDPHSGTPLSDHEALMATLFVRHSPPQQNPSSTHGPAERSPLMCVLKEAWTELGLGMAQARWWATFASYVIGLGLLLLALLCVLAAGGGAGEAAILLWTPSVGLVLWAGAFYLFHVQEVNGLYRAQAELQHVLGRAREAQDLGPEPQPALLLGQQEGDRTKEQ
Purification Method IgG purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6610
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.