missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSFL1C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31402
This item is not returnable.
View return policy
Description
NSFL1C Polyclonal specifically detects NSFL1C in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| NSFL1C | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| NSFL1C | |
| Recombinant protein encompassing a sequence within the center region of human NSFL1C. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| human NSFL1C protein. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| dJ776F14.1, NSFL1 (p97) cofactor (p47), NSFL1 cofactor p47, p47, p97 cofactor p47, SHP1 homolog, UBX domain protein 2C, UBX domain-containing protein 2C, UBX1, UBXD10, UBXN2CMGC3347 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 55968 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction