missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSFL1C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38307-100ul
This item is not returnable.
View return policy
Description
NSFL1C Polyclonal antibody specifically detects NSFL1C in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| NSFL1C | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells | |
| dJ776F14.1, NSFL1 (p97) cofactor (p47), NSFL1 cofactor p47, p47, p97 cofactor p47, SHP1 homolog, UBX domain protein 2C, UBX domain-containing protein 2C, UBX1, UBXD10, UBXN2CMGC3347 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-185 of human NSFL1C (NP_057227.2).,, Sequence:, MAAERQEALREFVAVTGAEEDRARFFLESAGWDLQIALASFYEDGGDEDIVTISQATPSSVSRGTAPSDNRVTSFRDLIHDQDEDEEEEEGQRFYAGGSERSGQQIVGPPRKKSPNELVDDLFKGAKEHGAVAVERVTKSPGETSKPRPFAGGGYRLGAAPEEESAYVAGEKRQHSSQDVHVVLK | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55968 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction