missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NSF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578095
This item is not returnable.
View return policy
Description
Positive Control: 293T, mouse brain, rat brain Predicted Reactivity: Xenopus laevis (88%), Pig (97%), Chicken (91%), Bovine (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Required for vesicle-mediated transport. Catalyzes the fusion of transport vesicles within the Golgi cisternae. Is also required for transport from the endoplasmic reticulum to the Golgi stack. Seem to function as a fusion protein required for the delivery of cargo proteins to all compartments of the Golgi stack independent of vesicle origin.
Specifications
| NSF | |
| Polyclonal | |
| Unconjugated | |
| NSF | |
| AI316878; AU020090; AU067812; Erg1; NEM-sensitive fusion protein; N-ethylmaleimide sensitive factor; N-ethylmaleimide sensitive factor, vesicle fusing ATPase; N-ethylmaleimide sensitive fusion protein; N-ethylmaleimide-sensitive factor; N-ethylmaleimide-sensitive factor-like protein; N-ethylmaleimide-sensitive fusion protein; NSF; Protein SKD2; Skd2; suppressor of K(+) transport growth defect 2; Vesicle-fusing ATPase; vesicle-fusing ATPase-like protein; vesicular-fusion protein NSF | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18195, 4905, 60355 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.27 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P46459, P46460, Q9QUL6 | |
| NSF | |
| Recombinant protein encompassing a sequence within the center region of human NSF. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction