Learn More
NSF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA129902
Description
PA1-29902 detects NSF from mouse, rat samples. PA1-29902 has been successfully used in immunohistochemistry (frozen tissue), immunoprecipitation, Western blot applications. The PA1-29902 immunogen is: Synthetic peptide corresponding to amino acids around 731 of rat NSF.
NSF (N-ethylmaleimide sensitive fusion protein) is a critical component of the SNARE (soluble NSF attachment protein receptors) protein complex that is involved in synaptic vesicle trafficking. Specifically, NSF has been found to be essential in membrane
Specifications
| NSF | |
| Polyclonal | |
| Unconjugated | |
| P46460, Q9QUL6 | |
| NSF | |
| Synthetic peptide corresponding to amino acids around 731 of rat NSF. | |
| 50 μg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
| 0.5 mg/mL | |
| Nsf | |
| N-ethylmaleimide sensitive fusion protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 18195, 60355 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.