missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NSF Polyclonal Antibody
GREENER_CHOICE

Product Code. 16516001
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16516001 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16516001 Supplier Invitrogen™ Supplier No. PA587525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: KHVKGILLYG PPGCGKTLLA RQIGKMLNAR EPKVVNGPEI LNKYVGESEA NIRKLFADAE EEQRRLGANS GLHIIIFDEI DAICKQRGSM AGSTGVHDTV VNQLLSKIDG VEQLNNILVI GMTNRPDLID EALLRPGRLE VKMEIGLPDE Positive Samples: Mouse brain, Rat brain; Cellular Location: Cytoplasm.

Required for vesicle-mediated transport. Catalyzes the fusion of transport vesicles within the Golgi cisternae. Is also required for transport from the endoplasmic reticulum to the Golgi stack. Seem to function as a fusion protein required for the delivery of cargo proteins to all compartments of the Golgi stack independent of vesicle origin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NSF
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene NSF
Gene Accession No. P46459, P46460, Q9QUL6
Gene Alias AI316878; AU020090; AU067812; Erg1; NEM-sensitive fusion protein; N-ethylmaleimide sensitive factor; N-ethylmaleimide sensitive factor, vesicle fusing ATPase; N-ethylmaleimide sensitive fusion protein; N-ethylmaleimide-sensitive factor; N-ethylmaleimide-sensitive factor-like protein; N-ethylmaleimide-sensitive fusion protein; NSF; Protein SKD2; Skd2; suppressor of K(+) transport growth defect 2; Vesicle-fusing ATPase; vesicle-fusing ATPase-like protein; vesicular-fusion protein NSF
Gene Symbols NSF
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 251-400 of human NSF (NP_0061692).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18195, 4905, 60355
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.