missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35086-100ul
This item is not returnable.
View return policy
Description
NSF Polyclonal antibody specifically detects NSF in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| NSF | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 0.5ug-4ug antibody for 400ug-600ug extracts of whole cells | |
| EC 3.6.4.6, NEM-sensitive fusion protein, N-ethylmaleimide-sensitive factor, N-ethylmaleimide-sensitive factor-like protein, N-ethylmaleimide-sensitive fusion protein, SKD2, vesicle-fusing ATPase, Vesicular-fusion protein NSF | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 251-400 of human NSF (NP_006169.2).,, Sequence:, KHVKGILLYGPPGCGKTLLARQIGKMLNAREPKVVNGPEILNKYVGESEANIRKLFADAEEEQRRLGANSGLHIIIFDEIDAICKQRGSMAGSTGVHDTVVNQLLSKIDGVEQLNNILVIGMTNRPDLIDEALLRPGRLEVKMEIGLPDE | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 4905 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction