missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nse2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76263
This item is not returnable.
View return policy
Description
Nse2 Polyclonal specifically detects Nse2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| Nse2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| C8orf36, E3 SUMO-protein ligase NSE2, EC 6.3.2, EC 6.3.2.-, FLJ32440, hMMS21, methyl methanesulfonate sensitivity gene 21, MMS21 homolog, MMS21chromosome 8 open reading frame 36, Non-SMC element 2 homolog, non-SMC element 2 homolog (MMS21, S. cerevisiae), non-SMC element 2, MMS21 homolog (S. cerevisiae), Non-structural maintenance of chromosomes element 2 homolog, NSE2 | |
| Rabbit | |
| 30 kDa | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 0.5 - 1 ug/ml., ELISA 1:100-1:2000 | |
| NP_775956 | |
| NSMCE2 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human MMS21. The immunogen is located within amino acids 170 - 220 of MMS21. Amino Acid Squence: CIVRMIESRQKRKKKA | |
| Affinity Purified | |
| RUO | |
| 286053 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction