Learn More
NSE Mouse anti-Human, CoraLite 488, Clone: 6F8G3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66150-100UL
Description
The NSE antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human NSE. This antibody recognizes human, mouse, rat, pig antigen. The NSE antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
| NSE | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 2026 | |
| IgG1 |
| Immunofluorescence | |
| 6F8G3 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |