missing translation for 'onlineSavingsMsg'
Learn More

NSDHL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£169.00 - £255.00

Specifications

Antigen NSDHL
Dilution Western Blot 1:500-1:1000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18697735
View Documents
Novus Biologicals
NBP2-99165-100ul
100 μg
£255.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18626245
View Documents
Novus Biologicals
NBP2-99165-50ul
50 μg
£169.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NSDHL Polyclonal antibody specifically detects NSDHL in Human samples. It is validated for Western Blot
Specifications

Specifications

NSDHL
Western Blot
Unconjugated
Rabbit
metabolism
0.2 um filtered solution in PBS
50814
IgG
Antigen and protein A Affinity-purified
Western Blot 1:500-1:1000
Polyclonal
Purified
RUO
Human
EC 1.1.1.170, H105e3, member 1, NAD(P) dependent steroid dehydrogenase-like, SDR31E1, sterol-4-alpha-carboxylate 3-dehydrogenase, decarboxylating, XAP104
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NSDHL
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.