missing translation for 'onlineSavingsMsg'
Learn More

NSD3 Rabbit anti-Human, Mouse, Rat, Clone: 3U4F9, Novus Biologicals™

Product Code. 18391256 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18391256 20 μg 20µL
18355495 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18391256 Supplier Novus Biologicals Supplier No. NBP31544920UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

NSD3 Monoclonal antibody specifically detects NSD3 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NSD3
Applications Western Blot
Classification Monoclonal
Clone 3U4F9
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 2.1.1.43, FLJ20353, histone-lysine N-methyltransferase NSD3, MGC126766, MGC142029, NSD3DKFZp667H044, Nuclear SET domain-containing protein 3, pp14328, Protein whistle, WHSC1-like 1 isoform 9 with methyltransferase activity to lysine, WHSC1-like protein 1, Wolf-Hirschhorn syndrome candidate 1-like 1, Wolf-Hirschhorn syndrome candidate 1-like protein 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human NSD3 (Q9BZ95).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 54904
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.