missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSD3 Antibody (7B4), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04324AF647
This item is not returnable.
View return policy
Description
NSD3 Monoclonal specifically detects NSD3 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| NSD3 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| WHSC1L1 | |
| Recombinant human NSD3 (383-660aa) purified from E. coli | |
| 0.1 mL | |
| Cancer | |
| 54904 | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| 7B4 | |
| Western Blot, ELISA | |
| EC 2.1.1.43, FLJ20353, histone-lysine N-methyltransferase NSD3, MGC126766, MGC142029, NSD3DKFZp667H044, Nuclear SET domain-containing protein 3, pp14328, Protein whistle, WHSC1-like 1 isoform 9 with methyltransferase activity to lysine, WHSC1-like protein 1, Wolf-Hirschhorn syndrome candidate 1-like 1, Wolf-Hirschhorn syndrome candidate 1-like protein 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human NSD3 mAb, clone 2E9, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human NSD3 protein. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction