missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NSD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553385
This item is not returnable.
View return policy
Description
Immunogen sequence: SANGKTPSCE VNRECSVFLS KAQLSSSLQE GVMQKFNGHD ALPFIPADKL KDLTSRVFNG EPGAHDAKLR FESQEMKGIG TPPNTTPIKN GSPEIKLKIT KTYMNGKPLF ESSICGD Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
This gene encodes a protein that contains four domains present in other developmental proteins: a PWWP domain, an HMG box, a SET domain, and a PHD-type zinc finger. It is expressed ubiquitously in early development. Wolf-Hirschhorn syndrome (WHS) is a malformation syndrome associated with a hemizygous deletion of the distal short arm of chromosome 4. This gene maps to the 165 kb WHS critical region and has also been involved in the chromosomal translocation t(4;14)(p16.3;q32.3) in multiple myelomas. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms. Some transcript variants are nonsense-mediated mRNA (NMD) decay candidates, hence not represented as reference sequences.
Specifications
| NSD2 | |
| Polyclonal | |
| Unconjugated | |
| NSD2 | |
| 5830445G22Rik; 9430010A17Rik; AW555663; C130020C13Rik; D030027O06Rik; D930023B08Rik; Histone-lysine N-methyltransferase NSD2; histone-lysine N-methyltransferase NSD2; LOW QUALITY PROTEIN: histone-lysine N-methyltransferase NSD2; IL5 promoter REII region-binding protein; Kiaa1090; LOW QUALITY PROTEIN: histone-lysine N-methyltransferase NSD2; mKIAA1090; MMSET; multiple myeloma SET domain containing protein type III; multiple myeloma SET domain-containing protein; NSD2; nuclear receptor binding SET domain protein 2; nuclear SET domain-containing protein 2; probable histone-lysine N-methyltransferase NSD2; Protein trithorax-5; putative histone-lysine N-methyltransferase NSD2-like protein; REIIBP; RGD1565590; trithorax/ash1-related protein 5; TRX5; WHS; Whsc1; Whsc1l; Wolf-Hirschhorn syndrome candidate 1; Wolf-Hirschhorn syndrome candidate 1 (human); Wolf-Hirschhorn syndrome candidate 1 protein; Wolf-Hirschhorn syndrome candidate 1 protein homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7468 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O96028 | |
| NSD2 | |
| Recombinant protein corresponding to Human NSD2. Recombinant protein control fragment (Product #RP-91616). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction