missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NSD1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15919774
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15919774 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15919774 Supplier Invitrogen™ Supplier No. PA567097

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: FSDVHFDSKVK QSDPGKISEK GLSFENGKGP ELDSVMNSEN DELNGVNQVV PKKRWQRLNQ RRTKPRKRMN RFKEKENSEC Highest antigen sequence identity to the following orthologs - mouse 80%, rat 72%.

This gene encodes a protein containing a SET domain, 2 LXXLL motifs, 3 nuclear translocation signals (NLSs), 4 plant homeodomain (PHD) finger regions, and a proline-rich region. The encoded protein enhances androgen receptor (AR) transactivation, and this enhancement can be increased further in the presence of other androgen receptor associated coregulators. This protein may act as a nucleus-localized, basic transcriptional factor and also as a bifunctional transcriptional regulator. Mutations of this gene have been associated with Sotos syndrome and Weaver syndrome. One version of childhood acute myeloid leukemia is the result of a cryptic translocation with the breakpoints occurring within nuclear receptor-binding Su-var, enhancer of zeste, and trithorax domain protein 1 on chromosome 5 and nucleoporin, 98-kd on chromosome 11. Two transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NSD1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NSD1
Gene Accession No. Q96L73
Gene Alias AI528500; Androgen receptor coactivator 267 kDa protein; androgen receptor-associated coregulator 267; androgen receptor-associated protein of 267 kDa; ARA267; H3-K36-HMTase; H4-K20-HMTase; Histone-lysine N-methyltransferase, H3 lysine-36 and H4 lysine-20 specific; KMT3B; Lysine N-methyltransferase 3B; NR-binding SET domain-containing protein; NSD1; nuclear receptor binding SET domain protein 1; nuclear receptor-binding SET domain-containing protein 1; nuclear receptor-binding SET-domain protein 1; SOTOS; SOTOS1; STO
Gene Symbols NSD1
Host Species Rabbit
Immunogen Recombinant Human NSD1. Recombinant protein control fragment (Product #RP-107743).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64324
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.