missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSD1, Mouse anti-Human, Clone: 3E6, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00064324-M06.100ug
This item is not returnable.
View return policy
Description
Sequence: DQTCELPRRNCLLPFSNPVNLDAPEDKDSPFGNGQSNFSEPLNGCTMQLSTVSGTSQNAYGQDSPSCYIPLRRLQDLASMINVEYLNGSADGSESFQDPEKSDSRAQTSpecifications
| NSD1 | |
| Monoclonal | |
| Unconjugated | |
| nuclear receptor binding SET domain protein 1 | |
| ARA267/DKFZp666C163/FLJ10684/FLJ22263/FLJ44628/KMT3B/SOTOS/STO | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3E6 | |
| In 1x PBS, pH 7.4 | |
| NM_022455 | |
| NSD1 | |
| NSD1 (NP_071900, 2 a.a. ∼ 109 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 64324 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction