missing translation for 'onlineSavingsMsg'
Learn More

NSC59984, MedChemExpress

Product Code. p-200102020
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16490328

Brand: Medchem Express HY19726

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NSC59984 induces mutant p53 protein degradation via MDM2 and the ubiquitin-proteasome pathway. NSC59984 acts by targeting GOF-mutant p53 and stimulates p73 to restore the p53 pathway signaling.

NSC59984 induces mutant p53 protein degradation via MDM2 and the ubiquitin-proteasome pathway. NSC59984 acts by targeting GOF-mutant p53 and stimulates p73 to restore the p53 pathway signaling.

Specifications

Chemical Name or Material NSC59984
CAS 803647-40-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C12H15N3O4
Quantity 5 mg
Solubility Information DMSO : 33.33 mg/mL (125.65 mM; Need ultrasonic)
SMILES O=C(N1CCN(C)CC1)/C=C/C2=CC=C([N+]([O-])=O)O2
Molecular Weight (g/mol) 265.27
Formula Weight 265.27
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.