missing translation for 'onlineSavingsMsg'
Learn More

NSC59984, MedChemExpress

Product Code. 16490328
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490328 5 mg 5mg
16457868 10 mg 10mg
16433817 50 mg 50mg
16427686 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16490328 Supplier Medchem Express Supplier No. HY19726
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress NSC59984 induces mutant p53 protein degradation via MDM2 and the ubiquitin-proteasome pathway. NSC59984 acts by targeting GOF-mutant p53 and stimulates p73 to restore the p53 pathway signaling.

NSC59984 induces mutant p53 protein degradation via MDM2 and the ubiquitin-proteasome pathway. NSC59984 acts by targeting GOF-mutant p53 and stimulates p73 to restore the p53 pathway signaling.

Specifications

Chemical Name or Material NSC59984
CAS 803647-40-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C12H15N3O4
Quantity 5 mg
Solubility Information DMSO : 33.33 mg/mL (125.65 mM; Need ultrasonic)
SMILES O=C(N1CCN(C)CC1)/C=C/C2=CC=C([N+]([O-])=O)O2
Molecular Weight (g/mol) 265.27
Formula Weight 265.27
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.