missing translation for 'onlineSavingsMsg'
Learn More

NSC5844, MedChemExpress

Product Code. 16485497
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485497 5 mg 5mg
16472758 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16485497 Supplier Medchem Express Supplier No. HY100033

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NSC5844 (RE-640) is a 4-aminoquinoline derivative, with antitumor and antimalarial activity.

NSC5844 (RE-640) is a 4-aminoquinoline derivative, with antitumor and antimalarial activity.

Specifications

Chemical Name or Material NSC5844
Physical Form Solid
CAS 140926-75-6
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H16Cl2N4
For Use With (Application) Cancer-programmed cell death
Synonym RE-640
Solubility Information DMSO : 1 mg/mL (2.61 mM; ultrasonic and warming and heat to 60°C)
SMILES ClC1=CC=C2C(NCCNC3=CC=NC4=CC(Cl)=CC=C34)=CC=NC2=C1
Molecular Weight (g/mol) 383.27
Formula Weight 383.27
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.