missing translation for 'onlineSavingsMsg'
Learn More

NSC319726, MedChemExpress

Product Code. 16460028
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460028 5 mg 5mg
16423577 50 mg 50mg
16407558 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16460028 Supplier Medchem Express Supplier No. HY18634
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress NSC319726 (ZMC1) is a mutant p53R175 reactivator; inhibits growth of fibroblasts expressing the p53R175 mutation (IC50 = 8 nM); shows no inhibition for p53 wild-type cells.

NSC319726 (ZMC1) is a mutant p53R175 reactivator

Specifications

Chemical Name or Material NSC319726
CAS 71555-25-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H14N4S
Quantity 5 mg
Synonym ZMC1
Solubility Information DMSO : 31.25 mg/mL (133.36 mM; Need ultrasonic)
SMILES C/C(C1=NC=CC=C1)=N\NC(N2CCC2)=S
Molecular Weight (g/mol) 234.32
Formula Weight 234.32
Percent Purity 98.06%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.