missing translation for 'onlineSavingsMsg'
Learn More

NSC232003, MedChemExpress

Product Code. 16466537
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16466537

Brand: Medchem Express HY103236

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NSC232003 is a highly potent and cell-permeable UHRF1 inhibitor, which inhibits DNA methylation in vitro and disrupts DNMT1/UHRF1 interactions at a cellular level.

NSC232003 is a highly potent and cell-permeable UHRF1 inhibitor, which inhibits DNA methylation in vitro and disrupts DNMT1/UHRF1 interactions at a cellular level.

Specifications

Chemical Name or Material NSC232003
Physical Form Solid
CAS 1905453-18-0
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C6H7N3O3
For Use With (Application) Cancer-programmed cell death
Solubility Information H2O : 2 mg/mL (11.82 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES O=C1NC(/C(C=N1)=C(NO)/C)=O
Molecular Weight (g/mol) 169.14
Formula Weight 169.14
Percent Purity 98.09%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.